Aluminiumsulfat Hydrat

Lieferant: Roth Carl
Gefahr

Synonyme: Aluminum(III) sulphate xhydrate

3731.1 3731.2
ROTH3731.1EA 0 EUR
ROTH3731.1 ROTH3731.2
Aluminiumsulfat Hydrat
Aluminiumsulfat Hydrat
Formel: Al₂(SO₄)₃·xH₂O
MDL Number: MFCD00149136
CAS-Nummer: 17927-65-0
EINECS: 233-135-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR