Indium(III)oxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indium trioxide, Diindium trioxide

M-Clarity™ quality level = MQ100

203424-25G 203424-5G
SIAL203424-25GEA 417 EUR
SIAL203424-25G SIAL203424-5G
Indium(III)oxid, Sigma-Aldrich®
Indium(III)oxid
Formel: In₂O₃
Schmelzpunkt: 715.05 °C
Dichte: 7.18 g/cm³ (20 °C)
MDL Number: MFCD00011060
CAS-Nummer: 1312-43-2
EINECS: 215-193-9
Merck Index: 12,04984

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR