Bariumtitanat ≥99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
Purity: 99%
CAS No.: 12047-27-7
Molecular Formula: BaTiO3
Formula Weight: 233.19
Physical Form: -325 Mesh Powder
Density: 5.85
Melting Point: 1620°
Solubility: Insoluble in water, alkalies. Slightly soluble in dilute acids. Soluble in concentrated acids
Merck Reference: 14,1000
MDL No.: MFCD00003447
Ferroelectric ceramics, single crystals, storage devices, dielectric amplifiers
Formel:
BaO₃Ti Schmelzpunkt: 1620 °C Dichte: 5.85 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00003447 CAS-Nummer: 12047-27-7 EINECS: 234-975-0 UN-Nummer: 1564 ADR: 6.1,III Merck Index: 14,1000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...