Bariumtitanat 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

196860025 196865000
ACRO196860025EA 196 EUR
ACRO196860025 ACRO196865000
Bariumtitanat 99%
Bariumtitanat
Formel: BaO₃Ti
Schmelzpunkt: 1625 °C
Dichte: 6.08 g/cm³
MDL Number: MFCD00003447
CAS-Nummer: 12047-27-7
EINECS: 234-975-0
ADR: 6.1,III
Merck Index: 12,01029

Bestellen

Spezifikation Testergebnisse

Appearance Off-white Powder
Assay 0.988 to 1.000 (BaO + SrO)/TiO2 Mole Ratio
Particle size 0.85 to 1.45 µm (Horiba, D50)
Silicon dioxide (SiO2) ≤0.02 %
Impurity ≤0.15 % (SrO)
Loss on ignition ≤0.5 %
Aluminium oxide (Al2O3) ≤0.04 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR