di-Kaliumthiosulfat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
12669-100G
12669-500G
SIAL12669-100GEA
96.8
EUR
SIAL12669-100G
SIAL12669-500G
di-Kaliumthiosulfat, Sigma-Aldrich®
di-Kaliumthiosulfat
Formel:
K₂O₃S₂ |
MDL Number:
MFCD00036292 CAS-Nummer: 10294-66-3 EINECS: 233-666-8 Merck Index: 12,07867 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...