1-(3,4-Xylyl)piperazin
Lieferant: Thermo Scientific
AC13696.1
AC13696.10
MAYBAC13696.1EA
0
EUR
MAYBAC13696.1
MAYBAC13696.10
1-(3,4-Xylyl)piperazin
1-(3,4-Xylyl)piperazin
Formel:
C₁₂H₁₈N₂ Siedepunkt: 135 °C (0,6 mmHg) Schmelzpunkt: 62 °C |
MDL Number:
MFCD00023129 CAS-Nummer: 1014-05-7 EINECS: 213-798-2 UN-Nummer: 3259 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...