Benzolsulfonamid, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ quality level = MQ100
108146-100G
108146-500G
108146-5G
SIAL108146-100GEA
35.7
EUR
SIAL108146-100G
SIAL108146-500G
SIAL108146-5G
Benzolsulfonamid, Sigma-Aldrich®
Benzolsulfonamid
Formel:
C₆H₇NO₂S Schmelzpunkt: 149…152 °C |
MDL Number:
MFCD00007930 CAS-Nummer: 98-10-2 EINECS: 202-637-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...