Methyl-2,4,6-trihydroxybenzoat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A12305.06 A12305.14 A12305.22
A12305.06EA 42.5 EUR
A12305.06 A12305.14 A12305.22
Methyl-2,4,6-trihydroxybenzoat ≥98%
Methyl-2,4,6-trihydroxybenzoat

Purity: 98% CAS No.: 3147-39-5 Molecular Formula: C8H8O5 Formula Weight: 184.15 Melting Point: 175-179° Beilstein Reference: 2837819

Formel: C₈H₈O₅
Schmelzpunkt: 175…179 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013969
CAS-Nummer: 3147-39-5
EINECS: 221-566-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR