2-Chlor-4-iodanilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A10742.14
A10742.06
A10742.22
A10742.14EA
177
EUR
A10742.14
A10742.06
A10742.22
2-Chlor-4-iodanilin ≥98%
2-Chlor-4-iodanilin
Purity: 98%
CAS No.: 42016-93-3
Molecular Formula: C6H5ClIN
Formula Weight: 253.47
Melting Point: 68-72°
Beilstein Reference: 2081106
UN No.: 2811
MDL No.: MFCD00025298
Formel:
C₆H₅ClIN Siedepunkt: 289 °C (1013 hPa) Schmelzpunkt: 68…72 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00025298 CAS-Nummer: 42016-93-3 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...