MES Natriumsalz , ULTROL® biologischer Puffer, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Sodium 2-Morpholinoethanesulphonate, Sodium 3-morpholin-1-ylethylsulphonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, MES-Na

475894-1 475894-100
475894-1EA 1060 EUR
475894-1 475894-100
MES Natriumsalz , ULTROL® biologischer Puffer, Millipore®
MES Natriumsalz

These Zwitterionic buffers are useful in the pH range of 5.5 to 6.7 and has a pKa of 6.10 at 25°C


Has an absorbance of ≤0.05 (100 mM, H2O, 260 nM).

Formel: C₆H₁₂NNaO₄S
Molekulargewicht: 217.22 g/mol
MDL Number: MFCD00065473
CAS-Nummer: 71119-23-8
EINECS: 275-203-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR