tert-Butylcarbamat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Carbamic acid tert-butyl ester
L00259.06
L00259.14
L00259.06EA
55.9
EUR
L00259.06
L00259.14
tert-Butylcarbamat ≥98%
tert-Butylcarbamat
Purity: 98+%
CAS No.: 4248-19-5
Molecular Formula: H2NCO2C(CH3)3
Formula Weight: 117.15
Melting Point: 106-110°
Beilstein Reference: 1744500
Formel:
C₅H₁₁NO₂ Schmelzpunkt: 106…110 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007962 CAS-Nummer: 4248-19-5 EINECS: 224-209-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...