Ethyldecanoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ethyl caprate, Capric acid ethyl ester, Decanoic acid ethyl ester, Decylic acid ethyl ester
M-Clarity™ quality level = MQ300
W243205-1KG-K
W243205-20KG-K
W243205-4KG-K
W243205-9KG-K
SIALW243205-1KG-KEA
108
EUR
SIALW243205-1KG-K
SIALW243205-20KG-K
SIALW243205-4KG-K
SIALW243205-9KG-K
Ethyldecanoat, Sigma-Aldrich®
Ethyldecanoat
Formel:
C₁₂H₂₄O₂ Siedepunkt: 245 °C (1013 hPa) Schmelzpunkt: –20 °C Dichte: 0.862 g/cm³ (20 °C) |
MDL Number:
MFCD00009581 CAS-Nummer: 110-38-3 EINECS: 203-761-9 Merck Index: 13,03811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...