Triacetin (Glycerintriacetat) ≥99%
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate
A17132.36
A17132.0E
A17132.36EA
31.6
EUR
A17132.36
A17132.0E
Triacetin (Glycerintriacetat) ≥99%
Triacetin (Glycerintriacetat)
Purity: 99%
CAS No.: 102-76-1
Molecular Formula: C9H14O6
Formula Weight: 218.21
Flash Point: 149°(300°F)
Physical Form: Liquid
Density: 1.155
Odour: Petroleum-like
Melting Point: 2-4°
Boiling Point: 257-259°
MDL No.: MFCD00008716
Formel:
C₉H₁₄O₆ Siedepunkt: 257…259 °C Schmelzpunkt: 2…4 °C Dichte: 1.155 Flammpunkt: 149 °C (300 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00008716 CAS-Nummer: 102-76-1 EINECS: 203-051-9 Merck Index: 13,09664 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...