Methylenblau Hydrat, rein
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methylthioninium chloride hydrate, Basic Blue 9, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride hydrate, C.I. 52015, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride xhydrate
414251000
414255000
414250025
ACRO414251000EA
61
EUR
ACRO414251000
ACRO414255000
ACRO414250025
Methylenblau Hydrat, rein
Methylenblau Hydrat
Formel:
C₁₆H₁₈ClN₃S·xH₂O Schmelzpunkt: 100…110 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150006 CAS-Nummer: 122965-43-9 EINECS: 200-515-2 |
Spezifikation Testergebnisse
Appearance | Green Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤22 % |
Titration with TiCl3 | ≥70 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...