L-Lysin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH

M-Clarity™ quality level = MQ200

L5501-25G L5501-100G L5501-10MG L5501-1G L5501-5G
SIALL5501-25GEA 185 EUR
SIALL5501-25G SIALL5501-100G SIALL5501-10MG SIALL5501-1G SIALL5501-5G
L-Lysin, Sigma-Aldrich®
L-Lysin
Formel: C₆H₁₄N₂O₂
Siedepunkt: 290 °C (1013 hPa)
Schmelzpunkt: 196 °C
MDL Number: MFCD00064433
CAS-Nummer: 56-87-1
EINECS: 200-294-2
Merck Index: 13,05656

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR