D(-)-Serin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-(-)-Serine, H-D-Ser-OH
M-Clarity™ quality level = MQ200
S4250-25G
S4250-5G
SIALS4250-25GEA
120
EUR
SIALS4250-25G
SIALS4250-5G
D(-)-Serin, Sigma-Aldrich®
D(-)-Serin
Formel:
C₃H₇NO₃ Schmelzpunkt: 235 °C |
MDL Number:
MFCD00004269 CAS-Nummer: 312-84-5 EINECS: 206-229-4 Merck Index: 12,08604 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...