Hexachloroplatinsäure Hydrat ≥99,999% (Metall-Basis), Premion®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Hydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate hydrate, Chloroplatinic acid hydrate, Dihydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate(IV) hydrate
Purity: 99.999%
CAS No.: 26023-84-7
Molecular Formula: H2PtCl6.xH2O
Formula Weight: 409.82(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Red-brown
Odour: Odourless
Melting Point: 60° dec.
Merck Reference: 14,7526
UN No.: 2507
MDL No.: MFCD00149909
Formel:
H₂Cl₆Pt·H₂O Schmelzpunkt: 60 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00149909 CAS-Nummer: 26023-84-7 EINECS: 241-010-7 UN-Nummer: 2507 ADR: 8,III Merck Index: 14,7526 |