Ethyl-(S)-(-)-lactat ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ethyl (S)-(-)-2-hydroxypropionate, Ethyl L(-)-lactate, (S)-Ethyl 2-hydroxypropanoate, (S)-(-)-Ethyl lactate
Purity: 99%
CAS No.: 687-47-8
Molecular Formula: C5H10O3
Formula Weight: 118.13
Flash Point: 46°(114°F)
Physical Form: Liquid
Density: 1.036
Refractive Index: 1.4130
Appearance: Colourless
Melting Point: -25°
Boiling Point: 152-155°
Beilstein Reference: 1720839
Merck Reference: 14,3817
Optical Rotation: -11° (neat)
UN No.: 1192
MDL No.: MFCD00065359
Formel:
C₅H₁₀O₃ Siedepunkt: 152…155 °C Schmelzpunkt: –25 °C Dichte: 1.036 Flammpunkt: 46 °C (114 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004518 CAS-Nummer: 687-47-8 EINECS: 211-694-1 UN-Nummer: 1192 ADR: 3,III Merck Index: 14,3817 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...