D(-)-Lysin Monohydrochlorid 99+%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-(-)-Lysine monohydrochloride, H-D-Lys-OH.HCl, D-Lysine hydrochloride, (R)-(-)-2,6-Diaminocaproic acid monohydrochloride, (R)-(-)-2,6-Diaminohexanoic acid monohydrochloride

243080010 243080100 243080500
ACRO243080010EA 33.6 EUR
ACRO243080010 ACRO243080100 ACRO243080500
D(-)-Lysin Monohydrochlorid 99+%
D(-)-Lysin Monohydrochlorid
Formel: C₆H₁₅ClN₂O₂
Schmelzpunkt: 266 °C (dec.)
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00012920
CAS-Nummer: 7274-88-6
EINECS: 230-691-6

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 3 h)
Sulfated ash ≤0.5 %
Specific optical rotation -19° to -22° (20°C, 589 nm) (c=5, 5 N HCl)
Titration with HClO4 99.0 to 101.0 %
Arsenic (As) ≤1 ppm
Iron (Fe) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR