Natriumdiethyldithiocarbamat Trihydrat ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate

33379.22 33379.36
33379.22EA 66.8 EUR
33379.22 33379.36
Natriumdiethyldithiocarbamat Trihydrat ACS
Natriumdiethyldithiocarbamat Trihydrat

CAS No.: 20624-25-3 Molecular Formula: (C2H5)2NCS2Na.3H2O Formula Weight: 225.31 (171.28anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White to off-white Melting Point: 94-102° Beilstein Reference: 3920507 Merck Reference: 14,3378 MDL No.: MFCD00150617

Formel: (C₂H₅)₂NCSSNa·3H₂O
Molekulargewicht: 225.31 g/mol
Schmelzpunkt: 94…102 °C
Dichte: 1.1 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00150617
CAS-Nummer: 20624-25-3
EINECS: 205-710-6
UN-Nummer: 3077
ADR: 9 III
Merck Index: 14,3378

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR