Kupfer(II)sulfat 0.1 mol/l, Titripur® volumetrische Lösung, Supelco®
Lieferant: Merck
Synonyme: Cupric sulphate
M-Clarity™ Qualitätsstufe = MQ100
1.02784.1000 1.02784.1003
1.02784.1000EA 59.7 EUR
1.02784.1000 1.02784.1003
Kupfer(II)sulfat 0.1 mol/l, Titripur® volumetrische Lösung, Supelco®
Kupfer(II)sulfat
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formel: CuSO₄
Molekulargewicht: 159.61 g/mol
Schmelzpunkt: 200
Dichte: 1.02 g/m3 (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00010981
CAS-Nummer: 7758-98-7
EINECS: 231-847-6
UN-Nummer: 3082
ADR: 9 III
Merck Index: 12,02722
Bestellen
Spezifikation Testergebnisse
Form liquid
Amount-of-substance concentration 0.0995 - 0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.