Calciumacetat xHydrat
Lieferant: Roth Carl
Synonyme:
Acetic acid calcium salt hydrate, Calcium di(acetate) hydrate, Calcium acetate hydrate
0260.1
0260.2
0260.3
ROTH0260.1EA
0
EUR
ROTH0260.1
ROTH0260.2
ROTH0260.3
Calciumacetat xHydrat
Calciumacetat xHydrat
Formel:
(H₃CCOO)₂Ca·nH₂O Molekulargewicht: 176.18 g/mol Dichte: 1.5 g/cm³ (20 °C) |
MDL Number:
MFCD00150019 CAS-Nummer: 114460-21-8 EINECS: 200-540-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...