Calciumacetat xHydrat FCC, DAC, reinst, SAFC®
Lieferant: MERCK PRODUCTION CHEMICALS
Synonyme: Acetic acid calcium salt hydrate, Calcium di(acetate) hydrate, Calcium acetate hydrate
1.02052.1000 1.02052.9029
1.02052.1000EA 213 EUR
1.02052.1000 1.02052.9029
Calciumacetat xHydrat FCC, DAC, reinst, SAFC®
Calciumacetat xHydrat
Formel: (H₃CCOO)₂Ca·nH₂O
Molekulargewicht: 176.18 g/mol
Dichte: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00150019
CAS-Nummer: 114460-21-8
EINECS: 200-540-9