Calciumacetat xHydrat ≥99,9965% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
Synonyme: Acetic acid calcium salt hydrate, Calcium di(acetate) hydrate, Calcium acetate hydrate
12974.06 12974.14
12974.06EA 67.7 EUR
12974.06 12974.14
Calciumacetat xHydrat ≥99,9965% (Metall-Basis), Puratronic®
Calciumacetat xHydrat
Purity: 99.9965% CAS No.: 114460-21-8 Molecular Formula: Ca(OOCCH3)2.xH2O Formula Weight: 158.17(anhy) Physical Form: Powder Density: 1.5 Melting Point: dec. Beilstein Reference: 3692527 Merck Reference: 14,1646 MDL No.: MFCD00012448
Formel: (H₃CCOO)₂Ca·nH₂O
Molekulargewicht: 176.18 g/mol
Schmelzpunkt: Zersetzung
Dichte: 1.5
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00150019
CAS-Nummer: 114460-21-8
EINECS: 200-540-9
Merck Index: 14,1646