DL-Lysin Monohydrat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: (±)-Lysine monohydrate, (±)-2,6-Diaminohexanoic acid monohydrate, (±)-2,6-Diaminocaproic acid monohydrate, H-DL-Lys-OH monohydrate

L07576.06 L07576.14
L07576.06EA 30.8 EUR
L07576.06 L07576.14
DL-Lysin Monohydrat ≥98%
DL-Lysin Monohydrat

Purity: 98+% CAS No.: 885701-25-7 Molecular Formula: C6H14N2O2.H2O Formula Weight: 164.21 (146.19anhy) Melting Point: ca 110° Beilstein Reference: 1616991

Formel: C₆H₁₄N₂O₂·1H₂O
Schmelzpunkt: ∼ 110 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00151036
CAS-Nummer: 885701-25-7
EINECS: 200-740-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR