(±)-Ethyl-2-methylbutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-2-Methylbutanoic acid ethyl ester, (±)-2-Methylbutyric acid ethyl ester, Ethyl 2-methylbutanoate

306886-100ML 306886-5ML
SIAL306886-100MLEA 51.6 EUR
SIAL306886-100ML SIAL306886-5ML
(±)-Ethyl-2-methylbutyrat, Sigma-Aldrich®
(±)-Ethyl-2-methylbutyrat
Formel: C₇H₁₄O₂
MDL Number: MFCD00012217
CAS-Nummer: 7452-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR