Natriumpolyphosphat (Grahamsches Salz), EMPLURA®, Supelco®
Lieferant: Merck
Synonyme: Metaphosphoric acid, sodium salt, Sodium hexametaphosphate, Sodium polymetaphosphate, Polyphosphate sodium salt, Graham′s salt, Sodium metaphosphate, SHMP
M-Clarity™ quality level = MQ200
1.06529.1000 1.06529.5000 1.06529.9050
1.06529.1000EA 52.8 EUR
1.06529.1000 1.06529.5000 1.06529.9050
Natriumpolyphosphat (Grahamsches Salz), EMPLURA®, Supelco®
Natriumpolyphosphat (Grahamsches Salz)
Formel: (NaO₃P)n
Dichte: 2.4 g/cm³ (20 °C)
MDL Number: MFCD00134118
CAS-Nummer: 50813-16-6
EINECS: 233-782-9
Merck Index: 12,08814