Cysteaminiumchlorid ≥95%, kristalliner Feststoff

Lieferant: Cayman Chemical
Gefahr

22193-10 22193-100 22193-25 22193-50
CAYM22193-10EA 43 EUR
CAYM22193-10 CAYM22193-100 CAYM22193-25 CAYM22193-50
Cysteaminiumchlorid ≥95%, kristalliner Feststoff
Cysteaminiumchlorid

Cysteamine is a stable aminothiol with radioprotective activities.


It reduces ionizing radiation-induced death and chromosomal damage in mice in a dose-dependent manner. Cysteamine binds rapidly and temporarily to plasma proteins upon administration and this activity is directly correlated to its radioprotective effects. In vitro, 0.1 mM cysteamine depletes 90% of free cystine from cystinotic fibroblasts. Formulations containing cysteamine have been used to treat nephropathic cystinosis and reduce glomerular deterioration in humans.


Bitte beachten This product is not for human or veterinary use.

Formel: C₂H₇NS·HCl
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00012904
CAS-Nummer: 156-57-0
UN-Nummer: 3335
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR