Chemical resistant coverall, chemsafe, type 3B, SAFE®
Lieferant: DACH SCHUTZBEKLEIDUNG
54/XL 54/M 54/XXL 54/XXXL
DACH54/XLEA 0 EUR
DACH54/XL DACH54/M DACH54/XXL DACH54/XXXL
Chemical resistant coverall, chemsafe, type 3B, SAFE®
Einweg-Kleidung
The ChemSafe coverall protects the user against almost all chemical hazards.
  • Two zippers for maximum protection
  • Fully liquid-tight
  • Reinforced knee area
  • Antistatic material
Multilayered, laminated material, additionally reinforced at the knees. Two zippers (a one-way on the outside and a two-way zipper on the inside) feature a self-adhesive cover, as well as sealed and taped seams to guarantee the coverall is liquid- and spray-tight.
Furthermore, the antistatic coverall protects against solid particles, spray mist, and infectious agents. Consequently, ChemSafe is used in the chemical industry, in labs, and in the medical sector.
Zertifizierungen: Meets the requirements for a chemical protective coverall Type 3B, 4B, 5B, 6B. Also complies with the European standards EN 14126, EN 1073-2 and 1149-5.