Amberlite® IRA-67, freie Base 500 - 750 μm Ionenaustauscherharz, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

476633-1KG 476633-100G
476633-1KGEA 851 EUR
476633-1KG 476633-100G
Amberlite® IRA-67, freie Base 500 - 750 μm Ionenaustauscherharz, Supelco®
Amberlite® IRA-67, free base

Amberlite IRA 67 is a weakly basic anion exchange resin with high capacity for large organics.

MDL Number: MFCD00145567
CAS-Nummer: 65899-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR