Industrial gloves, uvex athletic lite
Lieferant: UVEX
6002706 6002707 6002708 6002709 6002710 6002711
UVEX6002706EA 42.4 EUR
UVEX6002706 UVEX6002707 UVEX6002708 UVEX6002709 UVEX6002710 UVEX6002711
Industrial gloves, uvex athletic lite
Handschuhe
These lightweight knitted PA/elastan gloves with NBR foam coating on palm and fingertips are ideal for mechanical precision work.
  • Good mechanical abrasion resistance
  • Good grip in dry and slightly damp areas
  • Breathable
  • Very good tactile feel
  • Very good fit
  • Highly flexible
Areas of application: Precision assembly work, fine assembly work, inspection.
Zertifizierungen: EN 388:2016 (4132X)