Distillation Standards, Supelco®
Lieferant: Merck
CRMDIGO-250ML CRMUDIGO-250ML CRMUDIKR-250ML
SIALCRMDIGO-250MLEA 310 EUR
SIALCRMDIGO-250ML SIALCRMUDIGO-250ML SIALCRMUDIKR-250ML
Distillation Standards, Supelco®
Standards Mineralölindustrie-Standards
Distillation Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Distillation ASTM D86 and ISO 3405.
  • Manufactured in accordance with ASTM / EN or IP test method protocol
  • Certified by Round Robin Method by independent test laboratories.
  • Tested exclusively by ISO 17025 accredited laboratories for ISO 17034 CRM's
  • Ensures good interlaboratory correlation
  • Supplied in tamper evident UN compliant security pack
Zertifizierungen: These products are certified reference material (CRMs) certified according to ISO 17034 under UKAS accreditation.