Multi-component alcohol mixtures for calibration, Cerilliant®, Supelco®
Lieferant: Merck
A-056-1.2ML A-057-1.2ML A-061-1.2ML A-076-1.2ML
A-056-1.2MLEA 24.9 EUR
A-056-1.2ML A-057-1.2ML A-061-1.2ML A-076-1.2ML
Multi-component alcohol mixtures for calibration, Cerilliant®, Supelco®
Standards Organische Standards
Multi-component calibration kits containing acetone, ethanol, isopropanol, and methanol varying concentrations. These kits ares suitable for calibration of blood alcohol testing methods using GC Headspace.
  • SNAP-N-SPIKE®/SNAP-N-SHOOT®
Bitte beachten Storage temperature: 2 to 8 °C (refrigerator)
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60acbf907d9d7ad294112f8f83e49edd235dcf83&vid=60acbf907d9d7ad294112f8f83e49edd235dcf83&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10a3303115299e4969f0dfcd3f3fe20c6e857eec&vid=10a3303115299e4969f0dfcd3f3fe20c6e857eec&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f64b3dcc7b697d7f81a322203ade1621bc43017&vid=9f64b3dcc7b697d7f81a322203ade1621bc43017&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=007157a31ad9f1a96fd34716baeadefb61c7af4c&vid=007157a31ad9f1a96fd34716baeadefb61c7af4c&prid=web&clid=SAPDAM