Poly(diallyldimethylammonium chloride) [pDADMAC], M.W. 200,000-350,000 20% (w/v) in Wasser, Sigma-Aldrich®
Lieferant: Merck
Synonyme: Polyquaternium-6, N,N-Dimethyl-N-2-propenyl-2-propen-1-aminiumchloridehomopolymer, PolyDADMAC
M-Clarity™ quality level = MQ100
409022-1L 409022-25ML 409022-4L
SIAL409022-1LEA 59.6 EUR
SIAL409022-1L SIAL409022-25ML SIAL409022-4L
Poly(diallyldimethylammonium chloride) [pDADMAC], M.W. 200,000-350,000 20% (w/v) in Wasser, Sigma-Aldrich®
Poly(diallyldimethylammonium chloride) [pDADMAC]
Viscosity: 250-500 cP(25 °C, Brookfield)
Formel: (C₈H₁₆ClN)n
Dichte: 1.04 g/mL (25 °C)
MDL Number: MFCD00192409
CAS-Nummer: 26062-79-3