MitoPQ ≥98%

Lieferant: Cayman Chemical
Gefahr

Synonyme: MitoParaquat, 1-Methyl-1'-(10-(triphenylphosphonio)decyl)-[4,4'-bipyridine]-1,1'-diium iodide

18808-5 18808-25
CAYM18808-5EA 316 EUR
CAYM18808-5 CAYM18808-25
MitoPQ ≥98%
MitoPQ

MitoPQ is comprised of a triphenylphosphonium lipophilic cation conjugated to the redox cycler paraquat. Driven by membrane potential, it accumulates selectively in the mitochondrial matrix where it produces superoxide by redox cycling at the flavin site of complex I. Thus, MitoPQ selectively increases superoxide production within mitochondria and can be used as a tool either in cells or in vivo to investigate the role of mitochondrial superoxide in pathology and redox signaling.

Formel: C₃₉H₄₆N₂P·3I
Lagerungstemperatur: Gefrierschrank
CAS-Nummer: 1821370-28-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR