1-Methoxy-5-methylphenaziniummethylsulfat ≥98%, kristalliner Feststoff
Lieferant: Cayman Chemical
|
Achtung
|
1-Methoxy-5-methylphenazinium (methyl sulphate) is a photochemically stable mediator of electron transport between NADPH and various electron acceptors such as tetrazolium dyes. It can be used to visualise NAD-linked dehydrogenase activities.
Formel:
C₁₅H₁₆N₂O₅S Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00040648 CAS-Nummer: 65162-13-2 |
Spezifikation Testergebnisse
Purity | ≥98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...