Methacrylic anhydride ≥93,0% (durch GC), farblose Flüssigkeit, Sigma-Aldrich®
Lieferant: Merck
Gefahr
M-Clarity™ Qualitätsstufe = MQ200
276685-500ML 276685-100ML
SIAL276685-500MLEA 103 EUR
SIAL276685-500ML SIAL276685-100ML
Methacrylic anhydride ≥93,0% (durch GC), farblose Flüssigkeit, Sigma-Aldrich®
Methacrylic anhydride
The copolymerisation of methacrylic anhydride at 36.6 °C produces its polymer.
It can be prepared directly from the corresponding acids through the use of dehydrating agents and indirectly from the reaction of sodium salts of the acids with corresponding acid chlorides.
Formel: C₈H₁₀O₃
Siedepunkt: 87 °C/13 mmHg (Lit.)
Dichte: 1,035 g/cm³ (25 °C) (Lit.)
Flammpunkt: 84 °C
MDL Number: MFCD00008586
CAS-Nummer: 760-93-0
UN-Nummer: 3265
ADR: 8,II
Bestellen
Spezifikation Testergebnisse
Stabiliser 1800 - 2200 ppm Topanol A
Purity (GC Area%) ≥93.0%
Infrared Spectrum Conforms to Structure