Bottle-top dispensers, Propenser/Propenser Plus
Lieferant: Sartorius
LH-723060 LH-723061 LH-723062 LH-723063 LH-723064 LH-723065 LH-723070 LH-723071 LH-723072 LH-723073 LH-723074 LH-723075
BHITLH-723060EA 347 EUR
BHITLH-723060 BHITLH-723061 BHITLH-723062 BHITLH-723063 BHITLH-723064 BHITLH-723065 BHITLH-723070 BHITLH-723071 BHITLH-723072 BHITLH-723073 BHITLH-723074 BHITLH-723075
Bottle-top dispensers, Propenser/Propenser Plus
Flaschenaufsatz-Dispenser
Prospenser is high quality bottle-top dispenser with excellent chemical resistance. It is safe, convenient, and customisable, making it perfect for a wide range of dispensing needs.
  • Volume range from 0,2 to 60 ml
  • Extremely high chemical and thermal resistance due to ceramic piston, glass cylinder, and highly resistant fluoroplastic parts
  • Incremental volume setting ensures accurate and repeatable results
  • Smooth and light to operate
  • Easy to attach aspiration tube with a secure threaded connection
  • Fully autoclavable
In addition, Prospenser Plus has a media recirculation system that allows unused media to be recovered from the device and returned to the bottle, as well as a 360° rotating dispensing head and universal connector at the back of the unit for drying tubes and filters.
Zertifizierungen: ISO 8655
Lieferumfang: Wide range of adapters included to fit most common bottle sizes.