Kaliumperiodat, (Trockenmasse) ≥99,8% (Trockenmasse, RT), Puriss. p.a. ACS, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Potassium tetroxoiodate (VII), Potassium metaperiodate

60450-100G 60450-500G
60450-100GEA 104.77 EUR
60450-100G 60450-500G
Kaliumperiodat, (Trockenmasse) ≥99,8% (Trockenmasse, RT), Puriss. p.a. ACS, Fluka™
Kaliumperiodat
Formel: KlO₄
Schmelzpunkt: 582 °C
Dichte: 3.62 g/cm³ (20 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011407
CAS-Nummer: 7790-21-8
EINECS: 232-196-0
UN-Nummer: 3085
ADR: 5.1,I
Merck Index: 13,07739

Bestellen

Spezifikation Testergebnisse

Assay (excluding dried substance) 99.8 - 100.3 %
Loss on drying (110°C) Max. 0.1 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 1 ppm
Sodium (Na) Max. 2000 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Other halogens (as Cl) Max. 100 ppm
Sulfate (SO₄) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR