8270 Benzidines mix
Lieferant: Restek
The analysis of semivolatile organic pollutants in solid waste, soil, water, and air matrices using gas chromatography - mass spectrometry (GC - MS).
The compounds are Benzidine, 3,3' - dichlorobenzidine, 3,3' - dimethylbenzidine (o-tolidine).
Zertifizierungen: EPA