8270 Benzidines mix
Lieferant: Restek
31688 31852
REST31688EA 51 EUR
REST31688 REST31852
8270 Benzidines mix
Standards Organische Standards
The analysis of semivolatile organic pollutants in solid waste, soil, water, and air matrices using gas chromatography - mass spectrometry (GC - MS).
The compounds are Benzidine, 3,3' - dichlorobenzidine, 3,3' - dimethylbenzidine (o-tolidine).
Zertifizierungen: EPA