Nα-Fmoc-N-δ-trityl-L-glutamine ≥98,0% (durch HPLC), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Fmoc-Gln(Trt)-OH
47674-25G-F
47674-5G-F
47674-100G-F
47674-1KG-F
SIAL47674-25G-FEA
113
EUR
SIAL47674-25G-F
SIAL47674-5G-F
SIAL47674-100G-F
SIAL47674-1KG-F
Nα-Fmoc-N-δ-trityl-L-glutamine ≥98,0% (durch HPLC), Sigma-Aldrich®
N-alpha-Fmoc-N-delta-trityl-L-glutamine
Formel:
C₃₉H₃₄N₂O₅ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00077056 CAS-Nummer: 132327-80-1 |
Spezifikation Testergebnisse
Assay (HPLC) | ≥98.0% |
Optical Activity | [α]/D -14.0±1.5°, c = 1% in DMF |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...