Ethanolamin ≥99,5% (durch GC), AnalaR NORMAPUR® ACS analytisches Reagens
Lieferant: VWR Chemicals
Synonyme:
2-Aminoethanol, Monoethanolamine
85955.290EA
89.5
EUR
85955.290
85955.320
Ethanolamin ≥99,5% (durch GC), AnalaR NORMAPUR® ACS analytisches Reagens
Ethanolamin
Formel:
NH₂CH₂CH₂OH Molekulargewicht: 61.08 g/mol Siedepunkt: 171 Schmelzpunkt: 10.5 Flammpunkt: 93 |
MDL Number:
MFCD00008183 CAS-Nummer: 141-43-5 Merck Index: 13,03762 |
|
Spezifikation Testergebnisse
Assay (G.C.) | Min. 99.5% |
Identity (IR-spectrum) | Passes test |
Colour (Hazen) | Max. 15 |
Aluminium (Al) | Max. 0.00005% |
Boron (B) | Max. 0.000002% |
Barium (Ba) | Max. 0.00001% |
Cadmium (Cd) | Max. 0.000005% |
Calcium (Ca) | Max. 0.00005% |
Chromium (Cr) | Max. 0.000002% |
Cobalt (Co) | Max. 0.000002% |
Copper (Cu) | Max. 0.000002% |
Heavy metals (as Pb) | Max. 0.0005% |
Iron (Fe) | Max. 0.00001% |
Lead (Pb) | Max. 0.00001% |
Magnesium (Mg) | Max. 0.00001% |
Manganese (Mn) | Max. 0.000002% |
Nickel (Ni) | Max. 0.000002% |
Tin (Sn) | Max. 0.00001% |
Zinc (Zn) | Max. 0.00001% |
Diethanolamine (G.C.) | Max. 0.1% |
Triethanolamine (G.C.) | Max. 0.1% |
Water (K.F.) | Max. 0.2% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...