DMSO resistant peel heat seal
Lieferant: 4titude Ltd.
4TI-0586-DMSO 4TI-0585 4TI-0587
FTIT4TI-0586-DMSOEA 1290 EUR
FTIT4TI-0586-DMSO FTIT4TI-0585 FTIT4TI-0587
DMSO resistant peel heat seal
Versiegeln Mikrotiterplatten-Dichtungen
Solvent resistant heat sealing foil suitable for low and room temperature compound storage.
  • Peelable from PP and COC plates, forms a weld seal to PE plates
  • Temperature range –80 to +40 °C