2-Amino-3-brom-5-methylpyridin

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Bromo-5-methylpyridin-2-amine, 2-Amino-3-bromo-5-picoline, 3-Bromo-5-methyl-2-pyridylamine

XBX00111.1 XBX00111EA XBX00111.50
MAYBXBX00111.1EA 0 EUR
MAYBXBX00111.1 MAYBXBX00111EA MAYBXBX00111.50
2-Amino-3-brom-5-methylpyridin
2-Amino-3-brom-5-methylpyridin
Formel: C₆H₇BrN₂
MDL Number: MFCD00068231
CAS-Nummer: 17282-00-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR