3-Brom-4-methylpyridin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Bromo-4-picoline
L19622.06
L19622.14
L19622.06EA
133
EUR
L19622.06
L19622.14
3-Brom-4-methylpyridin ≥98%
3-Brom-4-methylpyridin
Purity: 98%
CAS No.: 3430-22-6
Molecular Formula: C6H6BrN
Formula Weight: 172.03
Flash Point: 79°(174°F)
Refractive Index: 1.5620
Boiling Point: 199-200°
Beilstein Reference: 878354
MDL No.: MFCD00082592
Formel:
C₆H₆BrN Siedepunkt: 199…200 °C Dichte: 1.549 Flammpunkt: 79 °C (174 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00082592 CAS-Nummer: 3430-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...