3-Iodpyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ quality level = MQ100
539368-1G
539368-5G
SIAL539368-1GEA
45.7
EUR
SIAL539368-1G
SIAL539368-5G
3-Iodpyridin, Sigma-Aldrich®
3-Iodpyridin
Formel:
C₅H₄IN Siedepunkt: 90…92 °C (14 mmHg) Schmelzpunkt: 53.5 °C |
MDL Number:
MFCD00023553 CAS-Nummer: 1120-90-7 EINECS: 214-322-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...