3,4-Pyridindicarbonsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,4-Pyridinedicarboxylic acid, Cinchomeronic acid, Pyridine-3,4-dicarboxylicacid

A14580.18 A14580.09 A14580.30
A14580.18EA 182 EUR
A14580.18 A14580.09 A14580.30
3,4-Pyridindicarbonsäure ≥98%
3,4-Pyridindicarbonsäure

Purity: 98% CAS No.: 490-11-9 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: ca 255° dec. Beilstein Reference: 137242 Merck Reference: 14,2283 MDL No.: MFCD00006392

Formel: C₇H₅NO₄
Siedepunkt: 529 °C (1013 hPa)
Schmelzpunkt: ∼ 255 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006392
CAS-Nummer: 490-11-9
EINECS: 207-705-4
Merck Index: 14,2283

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR