2,3-Pyridindicarbonsäure 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Quinolinic acid

131851000 131850250 131855000
ACRO131851000EA 64.6 EUR
ACRO131851000 ACRO131850250 ACRO131855000
2,3-Pyridindicarbonsäure 99%
2,3-Pyridindicarbonsäure
Formel: C₇H₅NO₄
Siedepunkt: 357 °C (1013 hPa)
Schmelzpunkt: 188…190 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006295
CAS-Nummer: 89-00-9
EINECS: 201-874-8
Merck Index: 12,08258

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
Phosphate (PO4) ≤1.0 ppm
Sulfate (SO4) ≤1.0 ppm
Silicate (as SiO2) ≤0.01 ppm
Heavy metals (as Pb) ≤0.01 ppm
KMnO4-reducing substance Passes test
Conductivity 2.0 x 10-6 ohm-1 cm-1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR