2-Amino-3,5-dichlorpyridin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,5-Dichloro-2-pyridylamine, 3,5-Dichloropyridin-2-amine
SPB06843.250
SPB06843DA
MAYBSPB06843.250EA
0
EUR
MAYBSPB06843.250
MAYBSPB06843DA
2-Amino-3,5-dichlorpyridin
2-Amino-3,5-dichlorpyridin
Formel:
C₅H₄Cl₂N₂ Schmelzpunkt: 79…83 °C |
MDL Number:
MFCD00006313 CAS-Nummer: 4214-74-8 EINECS: 224-143-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...